missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FMOC-L-α-Alanine monohydrate, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437610050
Specifications
FMOC-L-α-Alanine monohydrate | |
-20 | |
White | |
5g | |
97.5% min. (HPLC) | |
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate | |
CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O | |
329.36 | |
329.36 | |
95% |
Authentic | |
4.78 to 6.78% | |
146.8°C to 148.1°C | |
207291-76-7 | |
C18H17NO4·H2O | |
GAPWKFLOMOFHGO-MERQFXBCSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate | |
18528118 | |
Crystalline Powder |
Chemical Identifiers
207291-76-7 | |
329.36 | |
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate |
C18H17NO4·H2O | |
GAPWKFLOMOFHGO-MERQFXBCSA-N | |
18528118 | |
CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O |