Please login to your online account to display your discounted pricing

FMOC-L-α-Alanine monohydrate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material FMOC-L-α-Alanine monohydrate
Assay ≥97.5%
Assay Percent Range 95%
Molecular Formula C18H17NO4·H2O
Formula Weight 329.36
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-alpha-alanine monohydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC437610250

 
thermo fisher scientific
AC437610250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC437610050

 
thermo fisher scientific
AC437610050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 329.36
Quantity 25g
Color White
Specific Rotation -16° to -20° (20°C, 589nm) c=1, DMF
Water 4.78 to 6.78%
Molecular Formula C18H17NO4·H2O
CAS 207291-76-7
Infrared Spectrum Authentic
Chemical Name or Material FMOC-L-α-Alanine monohydrate
Assay Percent Range 95%
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-alpha-alanine monohydrate
Packaging Glass bottle