missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FMOC-L-α-Alanine monohydrate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437610250

 View more versions of this product

Catalog No. AC437610250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

FMOC-L-α-Alanine monohydrate
Glass bottle
4.78 to 6.78%
146.8°C to 148.1°C
207291-76-7
C18H17NO4·H2O
GAPWKFLOMOFHGO-MERQFXBCSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate
18528118
Crystalline Powder
Authentic
-20
White
25g
97.5% min. (HPLC)
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate
CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O
329.36
329.36
95%

Chemical Identifiers

207291-76-7
329.36
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate
C18H17NO4·H2O
GAPWKFLOMOFHGO-MERQFXBCSA-N
18528118
CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O
SDS Documents