missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FMOC isothiocyanate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438660050
Specifications
FMOC isothiocyanate | |
Authentic | |
44.0°C to 45.0°C | |
199915-38-3 | |
C16H11NO2S | |
DHMYULZVFHHEHE-UHFFFAOYSA-N | |
9H-fluoren-9-ylmethyl N-(sulfanylidenemethylidene)carbamate | |
4154759 | |
Liquid |
65.6 to 71.0 % (C) 4.8 to 5.2 % (N) | |
Yellow to Brown | |
5g | |
98% | |
9-fluorenylmethoxycarbonyl isothiocyanate, 9-fluorenylmethoxycarbonylisothiocyanate, 9-fluorenylmethyl isothiocyanatoformate, 9h-fluoren-9-ylmethyl n-carbothioylcarbamate, 9h-fluoren-9-ylmethyl n-sulfanylidenemethylidene carbamate, fluoren-9-ylmethyloxycarbonyl isothiocyanate, fmoc isothiocyanate, fmoc isothiocyanate chn, fmoc-isothiocyanate, n-fmoc-isothiocyanate | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N=C=S | |
281.34 | |
281.34 | |
97% |
Chemical Identifiers
199915-38-3 | |
281.34 | |
9-fluorenylmethoxycarbonyl isothiocyanate, 9-fluorenylmethoxycarbonylisothiocyanate, 9-fluorenylmethyl isothiocyanatoformate, 9h-fluoren-9-ylmethyl n-carbothioylcarbamate, 9h-fluoren-9-ylmethyl n-sulfanylidenemethylidene carbamate, fluoren-9-ylmethyloxycarbonyl isothiocyanate, fmoc isothiocyanate, fmoc isothiocyanate chn, fmoc-isothiocyanate, n-fmoc-isothiocyanate | |
9H-fluoren-9-ylmethyl N-(sulfanylidenemethylidene)carbamate |
C16H11NO2S | |
DHMYULZVFHHEHE-UHFFFAOYSA-N | |
4154759 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N=C=S |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger