missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FMOC-D-phenylalanine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 441710050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
FMOC-D-phenylalanine | |
Authentic | |
C24H21NO4 | |
+ 40.00 | |
SJVFAHZPLIXNDH-JOCHJYFZSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid | |
978332 | |
98% |
86123-10-6 | |
97.5% min. (HPLC) | |
5g | |
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid | |
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
387.43 | |
387.43 |
Chemical Identifiers
86123-10-6 | |
387.43 | |
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid |
C24H21NO4 | |
SJVFAHZPLIXNDH-JOCHJYFZSA-N | |
978332 | |
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only