Please login to your online account to display your discounted pricing

FMOC-Cys(tBu)-OH, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC431270250

 View more versions of this product

Catalog No. AC431270250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 399.51
Quantity 25g
Color White
Melting Point 136° to 140°C
Chemical Name or Material FMOC-Cys(tBu)-OH
Specific Rotation -21.0° to -23.0° (20°C, 589nm) (c=1, DMF)
Assay Percent Range 98%
Molecular Formula C22H25NO4S
MDL Number MFCD00037130
CAS 67436-13-9
Packaging Glass bottle
Synonym Nalpha-FMOC-S-tert-butyl-L-cysteine
Infrared Spectrum Authentic

Specifications

Chemical Name or Material FMOC-Cys(tBu)-OH
CAS 67436-13-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H25NO4S
Formula Weight 399.51
MDL Number MFCD00037130
Synonym Nalpha-FMOC-S-tert-butyl-L-cysteine