missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fmoc-Arg(Pbf)-OH 98.0+%, TCI America™
$98.79 - $350.44
Chemical Identifiers
| CAS | 154445-77-9 |
|---|---|
| Molecular Formula | C34H40N4O7S |
| Molecular Weight (g/mol) | 648.775 |
| MDL Number | MFCD00235804 |
| InChI Key | HNICLNKVURBTKV-NDEPHWFRSA-N |
| Synonym | fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4 |
| PubChem CID | 11354259 |
| IUPAC Name | (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
| SMILES | CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C |
Chemical Identifiers
| 154445-77-9 | |
| 648.775 | |
| HNICLNKVURBTKV-NDEPHWFRSA-N | |
| 11354259 | |
| CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C |
| C34H40N4O7S | |
| MFCD00235804 | |
| fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4 | |
| (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
Specifications
| 154445-77-9 | |
| C34H40N4O7S | |
| 5 g | |
| HNICLNKVURBTKV-NDEPHWFRSA-N | |
| (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid | |
| 11354259 | |
| ≥98.0% (HPLC,T) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl) |
| White | |
| MFCD00235804 | |
| fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4 | |
| CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C | |
| 648.775 | |
| 648.78 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer