Please login to your online account to display your discounted pricing

Fluvoxamine maleate, Tocris


5-HT re-uptake inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS103310

 View more versions of this product

Catalog No. 103310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 434.41
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material Fluvoxamine maleate
Molecular Formula C15H21F3N2O2·C4H4O4
Synonym (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate
Recommended Storage Desiccate at +4°C
CAS 61718-82-9

Specifications

Chemical Name or Material Fluvoxamine maleate
Assay Percent Range >99%
Molecular Formula C15H21F3N2O2·C4H4O4
Formula Weight 434.41
Synonym (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate
Accessories

Desiccate at +4°C