Please login to your online account to display your discounted pricing

Fluticasone propionate, Tocris

Selective high affinity glucocorticoid agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material Fluticasone propionate
Assay Percent Range >98%
Molecular Formula C25H31F3O5S
Formula Weight 500.57
Synonym (6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid fluoromethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

200750

 
r & d systems
RDS200750
50mg Each for $895.76

200710

 
r & d systems
RDS200710
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 500.57
Quantity 50mg
Chemical Name or Material Fluticasone propionate
Assay Percent Range >98%
Synonym (6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid fluoromethyl ester
Molecular Formula C25H31F3O5S
CAS 80474-14-2
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature