Flushing Oil
Essential for high vacuum pump maintenance.
Manufacturer:  Fisher Scientific 93060002
 View more versions of this product
Catalog No. S41456
Description
Description
  • Safely remove sediment and contamination from high vacuum pumps
  • One or two flushings clean residue and protect pump
  • Pump can be refilled immediately after flushing
  • 1 gal. size
Use for flushing only. Should not be used for high vacuum operation.
Specifications
Specifications
1 gal.