Please login to your online account to display your discounted pricing

Flupirtine maleate, Tocris

KV7 channel activator

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material Flupirtine maleate
Assay Percent Range >98%
Molecular Formula C15H17FN4O2·C4H4O4
Formula Weight 420.4
Synonym N-[2-Amino-6-[[4-fluorophenyl)methyl]amino]-3-pyridinyl]carbamic acid ethyl ester maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

286750

 
r & d systems
RDS286750
50mg Each for $585.69

286710

 
r & d systems
RDS286710
10mg Each for $139.08
Description & Specifications

Specifications

Formula Weight 420.4
Quantity 50mg
CAS 75507-68-5
Molecular Formula C15H17FN4O2·C4H4O4
Chemical Name or Material Flupirtine maleate
Synonym N-[2-Amino-6-[[4-fluorophenyl)methyl]amino]-3-pyridinyl]carbamic acid ethyl ester maleate
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C