Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluphenazine dihydrochloride, 98%

Chemical Identifiers

Chemical Name or Material Fluphenazine dihydrochloride
CAS 146-56-5
Molecular Formula C22H26F3N3OS·2HCl
Formula Weight 510.44
MDL Number MFCD00055212
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2084822

  SDS
alfa aesar™
B2084822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2084836

  SDS
alfa aesar™
B2084836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2084814

  SDS
alfa aesar™
B2084814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Fluphenazine dihydrochloride
Quantity 100g
CAS 146-56-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 98%
Molecular Formula C22H26F3N3OS·2HCl
Formula Weight 510.44
MDL Number MFCD00055212
UN Number UN2811

Applications
It is employed as a dopamine D1 receptor antagonist and a D1DR and D2DR inhibitor. Fluphenazine Dihydrochloride has been used to deliver Fluphenazine to biological systems in studies probing the effects and metabolic fates of this commonly used dopamine antagonist. It is also employed as an antipsychotic.

Solubility
Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.