missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluoxetine hydrochloride, 99%

A selective serotonin reuptake inhibitor

Manufacturer:  Alfa Aesar™ J61197MD

 View more versions of this product

Catalog No. AAJ61197MD



Description

Description

Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

Solubility
Soluble in dimethylsulfoxide and water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Fluoxetine hydrochloride
158°C to 159°C
56296-78-7
MFCD00214288
adofen, fluctin, flunirin, fluoxeren, fluoxetine hcl, fluoxetine hydrochloride, lovan, prozac, sarafem
GIYXAJPCNFJEHY-UHFFFAOYSA-N
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
62857
Powder
White
250mg
C17H19ClF3NO
14,4185
Soluble in dimethylsulfoxide and water.
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
345.79
345.79
99%

Chemical Identifiers

56296-78-7
345.79
GIYXAJPCNFJEHY-UHFFFAOYSA-N
62857
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
C17H19ClF3NO
MFCD00214288
adofen, fluctin, flunirin, fluoxeren, fluoxetine hcl, fluoxetine hydrochloride, lovan, prozac, sarafem
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c

H302-H315-H318

EINECSNumber : 260-101-2

RTECSNumber : UI4050000

TSCA : No

Recommended Storage : Ambient temperatures

Documents