Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluorotriethoxysilane, 95%

Chemical Identifiers

Chemical Name or Material Fluorotriethoxysilane
CAS 358-60-1
Molecular Formula C6H15FO3Si
Linear Formula (CH3CH2O)3SiF
Formula Weight 182.27
MDL Number MFCD00236719
UN Number UN2924
Beilstein 1700825
Synonym Triethoxyfluorosilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1665206

  SDS
alfa aesar™
L1665206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1665214

  SDS
alfa aesar™
L1665214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.27
Chemical Name or Material Fluorotriethoxysilane
Molecular Formula C6H15FO3Si
Linear Formula (CH3CH2O)3SiF
Beilstein 1700825
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 95%
MDL Number MFCD00236719
CAS 358-60-1
UN Number UN2924
Synonym Triethoxyfluorosilane
Boiling Point 134° to 135°C
Quantity 5g
Flash Point 19°C (66°F)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.