missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Fluoropore™ Membrane Filter for Filtration of Solvents

Catalog No. FTHP04750
Change view
Click to view available options
Pore Size:
3 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
FTHP04750 3 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FTHP04750 Supplier MilliporeSigma™ Supplier No. FTHP04750
Only null left

The Fluoropore™ membrane filter is a hydrophobic, polytetrafluoroethylene (PTFE) polymer membrane bonded to a high density polyethylene support to improve filter handling characteristics for normal applications.

  • Fluoropore™ membranes provide broad chemical compatibility, high flow rates, and consistency
  • Catalog numbers containing (FHUP) are unlaminated, or produced without a backing
  • Applications: Clarifying acids, Bases and solvents (i.e. isopropanol and ethanol), Filtering and venting gases, UV Spectroscopy
  • Omnipore™ membranes are hydrophilic PTFE membranes compatible with virtually all solvents, acids, and alkaline solutions
    • Applications: Filtration of aqueous solutions, Clarifying acids and alkaline solutions
  • Features & Benefits:
    • Biologically and chemically inert
    • High porosity yields high flow rates
    • Hydrophobic and hydrophilic PTFE varieties for filtration of aqueous and organic-based samples

Specifications

Autoclavable Autoclavable
Filter Type Screen Filter
Material PTFE
Pore Size 3 μm
Sterility Non-sterile
Hydrophobicity Hydrophobic
Membrane Type Screen Filter
Type Screen Filter
Wettability Hydrophobic
Diameter (Metric) 47 mm