missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoromethyl Phenyl Sulfone, TCI America™

Manufacturer:  TCI America F03415G

 View more versions of this product

Catalog No. F03415G



Specifications

Specifications

Fluoromethyl Phenyl Sulfone
53°C
20808-12-2
MFCD00191650
DENHPZASLKBBHA-UHFFFAOYSA-N
fluoromethylsulfonylbenzene
2759036
Crystal-Powder at 20°C
White-Yellow
5g
C7H7FO2S
Fluoromethylsulfonylbenzene
C1=CC=C(C=C1)S(=O)(=O)CF
174.189
174.19
≥98.0% (GC)

Chemical Identifiers

20808-12-2
174.189
DENHPZASLKBBHA-UHFFFAOYSA-N
2759036
C1=CC=C(C=C1)S(=O)(=O)CF
C7H7FO2S
MFCD00191650
Fluoromethylsulfonylbenzene
fluoromethylsulfonylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents