missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%

$91.14 - $309.96

Chemical Identifiers

CAS 28523-86-6
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.056
MDL Number MFCD00153189
InChI Key DFEYYRMXOJXZRJ-UHFFFAOYSA-N
Synonym sevoflurane, sevofluran, ultane, 1,1,1,3,3,3-hexafluoro-2-fluoromethoxy propane, sevoflurano, sevofluranum, sevorane, sojourn, mr6s4, sevofluranum inn-latin
PubChem CID 5206
ChEBI CHEBI:9130
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
SMILES C(OC(C(F)(F)F)C(F)(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1689103 View Documents Alfa Aesar™
L1689103
1g Each for $91.14
 
AAL1689106 View Documents Alfa Aesar™
L1689106
5g Each for $309.96
 
Description

Description

Applications
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

28523-86-6
200.056
DFEYYRMXOJXZRJ-UHFFFAOYSA-N
5206
1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
C4H3F7O
MFCD00153189
sevoflurane, sevofluran, ultane, 1,1,1,3,3,3-hexafluoro-2-fluoromethoxy propane, sevoflurano, sevofluranum, sevorane, sojourn, mr6s4, sevofluranum inn-latin
CHEBI:9130
C(OC(C(F)(F)F)C(F)(F)F)F

Specifications

58°C
28523-86-6
MFCD00153189
14,8475
DFEYYRMXOJXZRJ-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
5206
200.06
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether
1g
C4H3F7O
2041023
sevoflurane, sevofluran, ultane, 1,1,1,3,3,3-hexafluoro-2-fluoromethoxy propane, sevoflurano, sevofluranum, sevorane, sojourn, mr6s4, sevofluranum inn-latin
C(OC(C(F)(F)F)C(F)(F)F)F
200.056
CHEBI:9130
≥98%
Safety and Handling

Safety and Handling

RTECSNumber : KO0737000