Please login to your online account to display your discounted pricing

Fluorodiphenylmethylsilane, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material Fluorodiphenylmethylsilane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H13FSi
Linear Formula (C6H5)2Si(F)CH3
Formula Weight 216.33
MDL Number MFCD03093773
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382910010

  SDS
thermo fisher scientific
AC382910010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 216.33
Quantity 1g
Color Clear light yellow
Packaging Glass bottle
Linear Formula (C6H5)2Si(F)CH3
Infrared Spectrum Authentic
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C13H13FSi
Refractive Index 1.5470 to 1.5500 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
CAS 17739-53-6
Chemical Name or Material Fluorodiphenylmethylsilane
MDL Number MFCD03093773
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.