Please login to your online account to display your discounted pricing

Fluorodimethylphenylsilane 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC382900010

 View more versions of this product

Catalog No. AC382900010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 154.26
Quantity 1g
Color Clear colorless
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Fluorodimethylphenylsilane
Assay Percent Range 97%
Molecular Formula C8H11FSi
Linear Formula (CH3)2Si(F)C6H5
MDL Number MFCD03093772
CAS 454-57-9
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Fluorodimethylphenylsilane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H11FSi
Linear Formula (CH3)2Si(F)C6H5
Formula Weight 154.26
MDL Number MFCD03093772
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

SDS & Safety Documents