Please login to your online account to display your discounted pricing

Fluorodimethylphenylsilane 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Fluorodimethylphenylsilane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H11FSi
Linear Formula (CH3)2Si(F)C6H5
Formula Weight 154.26
MDL Number MFCD03093772
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382900010

  SDS
thermo fisher scientific
AC382900010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC382900050

  SDS
thermo fisher scientific
AC382900050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 154.26
Physical Form Liquid
Quantity 1g
Color Clear colorless
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.
MDL Number MFCD03093772
Chemical Name or Material Fluorodimethylphenylsilane
Linear Formula (CH3)2Si(F)C6H5
Molecular Formula C8H11FSi
CAS 454-57-9
Assay Percent Range 97%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.