Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluorocyclohexane, 97%

Chemical Identifiers

Chemical Name or Material Fluorocyclohexane
CAS 372-46-3
Molecular Formula C6H11F
Formula Weight 102.15
MDL Number MFCD00021285
UN Number UN1993
Beilstein 1900795
Synonym Cyclohexyl fluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1301506

 
alfa aesar™
L1301506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.15
Chemical Name or Material Fluorocyclohexane
Assay 97%
Molecular Formula C6H11F
MDL Number MFCD00021285
CAS 372-46-3
UN Number UN1993
Beilstein 1900795
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 103° to 105°C
TSCA TSCA
Synonym Cyclohexyl fluoride
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Flash Point 5°C (41°F)
Melting Point 13°C
Quantity 5g

Applications
Fluorocyclohexane is used as an intermediate in chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.