Learn More
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals AC196700025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Fluoroboric acid | |
-90.0°C | |
130.0°C | |
Plastic bottle | |
2.5 kg | |
Adhering Crystals and/or Chunks | |
MFCD00011345 | |
01, 394 | |
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq | |
ODGCEQLVLXJUCC-UHFFFAOYSA-O | |
hydron;tetrafluoroborate | |
28118 | |
87.81 |
White | |
16872-11-0,7732-18-5,10043-35-3 | |
1.4100g/mL | |
1.41 | |
BF4H | |
HBF4 | |
1775 | |
15, 4180 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol | |
[H+].[B-](F)(F)(F)F | |
87.812 | |
CHEBI:38902 | |
48 to 51% (Acid) |
Chemical Identifiers
16872-11-0 | |
87.812 | |
ODGCEQLVLXJUCC-UHFFFAOYSA-O | |
28118 | |
hydron;tetrafluoroborate |
BF4H | |
MFCD00011345 | |
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq | |
CHEBI:38902 | |
[H+].[B-](F)(F)(F)F |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 240-898-3
RTECSNumber : ED2685000
TSCA : TSCA
Recommended Storage : May develop some turbidity or precipitate
RUO – Research Use Only