missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluorobenzene 99.0+%, TCI America™
Supplier: TCI America F0034100G
Specifications
Fluorobenzene | |
Colorless | |
C6H5F | |
100 g | |
benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene | |
FC1=CC=CC=C1 | |
96.10 | |
CHEBI:5115 | |
≥99.0% (GC) |
462-06-6 | |
85°C | |
MFCD00000280 | |
2387 | |
PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
fluorobenzene | |
10008 | |
96.10 | |
Liquid |
Chemical Identifiers
462-06-6 | |
96.10 | |
PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
10008 | |
fluorobenzene |
C6H5F | |
MFCD00000280 | |
benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene | |
CHEBI:5115 | |
FC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (9)-1232
RTECSNumber : DA0800000
TSCA : Yes