missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluorobenzene, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 119345000
Specifications
Fluorobenzene | |
−15°C | |
Glass bottle | |
1.02 | |
85.0°C | |
-42.0°C | |
462-06-6 | |
C6H5F | |
05,IV,632 | |
1,2,3,4,5-pentadeuterio-6-fluorobenzene, 4-fluorobenzene, benzene, fluoro, fluoranylbenzene, fluorbenzol, fluoro-benzene, fluorobenzenes, monofluorobenzene, phenyl fluoride, unii-g3tsz68k12 | |
PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
fluorobenzene | |
10008 | |
96.1 | |
99% |
1.0200g/mL | |
Authentic | |
1.4643 to 1.4663 | |
0.1% Max. (K.F.) | |
Colorless | |
500g | |
98.5% min. (GC) | |
C6H5F | |
15,4201 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol and water | |
C1=CC=C(C=C1)F | |
96.1 | |
CHEBI:5115 | |
Liquid |
Chemical Identifiers
462-06-6 | |
96.1 | |
1,2,3,4,5-pentadeuterio-6-fluorobenzene, 4-fluorobenzene, benzene, fluoro, fluoranylbenzene, fluorbenzol, fluoro-benzene, fluorobenzenes, monofluorobenzene, phenyl fluoride, unii-g3tsz68k12 | |
CHEBI:5115 | |
C1=CC=C(C=C1)F |
C6H5F | |
PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
10008 | |
fluorobenzene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
GHS Signal Word: Danger
EINECSNumber : 207-321-7