missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoroacetone, 97%, ACROS Organics™

$88.31 - $88.31

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C3H5FO
Linear Formula CH3COCH2F
Formula Weight 76.06
MDL Number MFCD00000451
Chemical Name or Material Fluoroacetone
Assay ≥96.0% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC151780010 SDS Thermo Fisher Scientific
AC151780010
Glass bottle 1g Each for $88.31
AC151780050 SDS Thermo Fisher Scientific
AC151780050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
CH3COCH2F
MFCD00000451
≥96.0% (GC)
C3H5FO
76.06
Fluoroacetone

Specifications

Red to dark red
430-51-3
Authentic
1.3660 to 1.3700 (20°C, 589nm)
97%
CH3COCH2F
MFCD00000451
≥96.0% (GC)
1g
1.0540g/mL
Glass bottle
Powder
C3H5FO
76.06
Fluoroacetone

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger