missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 164298-23-1 |
---|---|
Molecular Formula | C5H12FN2·F6P |
Molecular Weight (g/mol) | 264.13 |
MDL Number | MFCD02684443 |
InChI Key | ZAVXOOLKAGPJPI-UHFFFAOYSA-N |
Synonym | tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate |
PubChem CID | 2774761 |
IUPAC Name | [dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate |
SMILES | CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC434960250
|
Thermo Scientific Chemicals
434960250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentChemical Identifiers
164298-23-1 | |
264.13 | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
2774761 | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F |
C5H12FN2·F6P | |
MFCD02684443 | |
tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate |
Specifications
164298-23-1 | |
98.5% min. (HPLC) | |
C5H12FN2·F6P | |
25g | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate | |
2774761 | |
99% |
Authentic | |
Glass bottle | |
MFCD02684443 | |
tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F | |
264.13 | |
264.13 | |
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only