missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC434960010
Description
Coupling reagentSpecifications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate | |
Glass bottle | |
107°C to 110°C | |
164298-23-1 | |
C5H12FN2·F6P | |
acmc-209dqu, c5h12fn2.f6p, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, tetramethyl fluoroformamidium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F | |
264.13 | |
264.13 | |
99% |
Authentic | |
Yellow | |
1g | |
98.5% min. (HPLC) | |
MFCD02684443 | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate | |
2774761 | |
Powder |
Chemical Identifiers
164298-23-1 | |
264.13 | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
2774761 | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F |
C5H12FN2·F6P | |
MFCD02684443 | |
acmc-209dqu, c5h12fn2.f6p, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, tetramethyl fluoroformamidium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning