Learn More
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 99%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC434960050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentSpecifications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate | |
Yellow | |
98.5% min. (HPLC) | |
C5H12FN2·F6P | |
Glass bottle | |
5g | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F | |
264.13 | |
264.13 | |
99% |
164298-23-1 | |
107°C to 110°C | |
Authentic | |
MFCD02684443 | |
acmc-209dqu, c5h12fn2.f6p, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, tetramethyl fluoroformamidium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate | |
2774761 | |
Powder |
Chemical Identifiers
164298-23-1 | |
264.13 | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
2774761 | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F |
C5H12FN2·F6P | |
MFCD02684443 | |
acmc-209dqu, c5h12fn2.f6p, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, tetramethyl fluoroformamidium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only