Learn More
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5491006
Description
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate | |
104°C to 111°C | |
MFCD02684443 | |
tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate | |
2774761 | |
98% |
164298-23-1 | |
C5H12F7N2P | |
5 g | |
Soluble in water. | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F | |
264.128 | |
264.12 |
Chemical Identifiers
164298-23-1 | |
264.128 | |
ZAVXOOLKAGPJPI-UHFFFAOYSA-N | |
2774761 | |
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F |
C5H12F7N2P | |
MFCD02684443 | |
tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate | |
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only