missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%

Manufacturer:  Alfa Aesar™ H5491014

 View more versions of this product

Catalog No. AAH5491014



Description

Description

Applications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
25g
C5H12F7N2P
acmc-209dqu
ZAVXOOLKAGPJPI-UHFFFAOYSA-N
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate
2774761
98%
104°C to 111°C
164298-23-1
MFCD02684443
Soluble in water.
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F
264.128
264.12

Chemical Identifiers

164298-23-1
264.128
ZAVXOOLKAGPJPI-UHFFFAOYSA-N
2774761
CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F
C5H12F7N2P
MFCD02684443
acmc-209dqu
[dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents