Please login to your online account to display your discounted pricing

Fluoride Standard, Certified, 5.00ppm ±0.10ppm (1mL = 0.005mg F-), LabChem™

$47.83 - $47.83

Chemical Identifiers

Chemical Name or Material Fluoride Standard
CAS 7732-18-5
CAS Max % 99.999
CAS 7681-49-4
CAS Max % 0.001
Name Note 5ppm (1mL = 0.005mg F-)
Purity Grade Certified Grade
Linear Formula NaF
Formula Weight 41.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC145651

 
lab chem
LC145651
Poly Bottle 500mL Each for $47.83
Description & Specifications

Specifications

Formula Weight 41.99
Purity Grade Certified Grade
Physical Form Liquid
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Concentration 5.00ppm ±0.10ppm (1mL = 0.005mg F-)
CAS Max % 99.999
Solubility Soluble in water
Name Note 5ppm (1mL = 0.005mg F-)
Recommended Storage Room Temperature
Quantity 500mL
CAS 7732-18-5
Identification Passes Test
Traceability to NIST Traceable to NIST
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS 7681-49-4
Color Clear, colorless
Chemical Name or Material Fluoride Standard
CAS Max % 0.001
Packaging Poly Bottle
Linear Formula NaF
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature