Please login to your online account to display your discounted pricing

Fluoride Standard, Certified, 100.0ppm ±0.5ppm (1mL = 0.1mg F-), LabChem™

$49.71 - $49.71

Chemical Identifiers

Chemical Name or Material Fluoride Standard
CAS 7732-18-5
CAS Max % 99.98
CAS 7681-49-4
CAS Max % 0.02
Name Note 100ppm (1mL = 0.1mg F-)
Purity Grade Certified Grade
Linear Formula NaF
Formula Weight 41.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC145801

 
lab chem
LC145801
Poly Bottle 500mL Each for $49.71
Description & Specifications

Specifications

Formula Weight 41.99
Purity Grade Certified Grade
Solubility Soluble in water
Quantity 500mL
Recommended Storage Room Temperature
Name Note 100ppm (1mL = 0.1mg F-)
CAS 7732-18-5
Concentration 100.0ppm ±0.5ppm (1mL = 0.1mg F-)
Traceability to NIST Traceable to NIST
Physical Form Liquid
CAS Max % 0.02
Color Clear, colorless
Packaging Poly Bottle
CAS Max % 99.98
Chemical Name or Material Fluoride Standard
Identification Passes Test
CAS 7681-49-4
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Linear Formula NaF
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature