Please login to your online account to display your discounted pricing

Fluoride Standard, Certified, 10.0ppm ±0.1ppm (1mL = 0.01mg F-), LabChem™

$50.46 - $50.46

Chemical Identifiers

Chemical Name or Material Fluoride Standard
CAS 7732-18-5
CAS Max % 99.998
CAS 7681-49-4
CAS Max % 0.002
Name Note 10ppm (1mL = 0.01mg F-)
Purity Grade Certified Grade
Linear Formula NaF
Formula Weight 41.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC145701

 
lab chem
LC145701
Poly Bottle 500mL Each for $50.46
Description & Specifications

Specifications

Formula Weight 41.99
Purity Grade Certified Grade
Packaging Poly Bottle
Quantity 500mL
CAS Max % 99.998
CAS 7681-49-4
Recommended Storage Room Temperature
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Concentration 10.0ppm ±0.1ppm (1mL = 0.01mg F-)
Color Clear, colorless
Identification Passes Test
CAS 7732-18-5
Solubility Soluble in water
CAS Max % 0.002
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Chemical Name or Material Fluoride Standard
Name Note 10ppm (1mL = 0.01mg F-)
Physical Form Liquid
Linear Formula NaF
Traceability to NIST Traceable to NIST
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature