Please login to your online account to display your discounted pricing

Fluoride Standard, Certified, 0.50ppm ±0.03ppm (1mL = 0.0005mg F-), LabChem™

$40.54 - $40.54

Chemical Identifiers

Chemical Name or Material Fluoride Standard
CAS 7732-18-5
CAS Max % 99.9999
CAS 7681-49-4
CAS Max % 0.0001
Name Note 0.5ppm (1mL = 0.0005mg F-)
Purity Grade Certified Grade
Linear Formula NaF
Formula Weight 41.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC145551

 
lab chem
LC145551
Poly Bottle 500mL Each for $40.54
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 41.99
Chemical Name or Material Fluoride Standard
Color Clear, colorless
Quantity 500mL
Name Note 0.5ppm (1mL = 0.0005mg F-)
Physical Form Liquid
Concentration 0.50ppm ±0.03ppm (1mL = 0.0005mg F-)
Linear Formula NaF
CAS 7732-18-5
Identification Passes Test
CAS Max % 99.9999
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7681-49-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.0001
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature