missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 1000 ppm F-, Ricca Chemical
Supplier: Ricca Chemical Company RI31734
Specifications
Fluoride Standard | |
Natural Poly Bottle | |
98.0,0.22 | |
Colorless | |
FNa | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium fluoride | |
5235 | |
1mL = 1mg F- |
7681-49-4,7732-18-5 | |
120 mL | |
100.0,0.23 | |
1000ppm F- | |
MFCD00003524 | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
Liquid |
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
CHEBI:28741 | |
[F-].[Na+] |
FNa | |
MFCD00003524 | |
5235 | |
sodium fluoride |