Please login to your online account to display your discounted pricing

Fluoride Standard, 1mL = 0.01mg F-, 10ppm F-, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Fluoride Standard
Name Note 1mL = 0.01mg F-, 10ppm F-
CAS 7681-49-4
CAS 7732-18-5
Assay Percent Range 10.00 ± 0.05ppm
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R3170000120

 
ricca chemical company
RIR3170000120A
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R31700004A

 
ricca chemical company
RIR31700004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 120mL
CAS 7681-49-4
Chemical Name or Material Fluoride Standard
Name Note 1mL = 0.01mg F-, 10ppm F-
Assay Percent Range 10.00 ± 0.05ppm
Odor Odorless
CAS 7732-18-5
Packaging Poly natural
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution