missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 1mL = 0.005mg F, in TISAB Buffer (Color Coded Blue), Ricca Chemical
Manufacturer: Ricca Chemical Company RI316416
Specifications
Fluoride Standard | |
7681-49-4,1310-73-2,64-19-7,7647-14-5,13291-61-7,7732-18-5 | |
97.0,2.0,1.68,1.63,0.11,4.0E-4 | |
Poly natural | |
Blue |
5ppm F- | |
93.0,1.92,1.61,1.56,0.11,4.0E-4 | |
Liquid | |
1 mL = 0.005 mg F- | |
500mL |