Please login to your online account to display your discounted pricing

Fluoride Standard, 1mL = 0.005mg F, in TISAB Buffer (Color Coded Blue), Ricca Chemical

$107.48 - $107.48

Chemical Identifiers

Chemical Name or Material Fluoride Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 1310-73-2
CAS Min % 1.92
CAS Max % 2.0
CAS 64-19-7
CAS Min % 1.61
CAS Max % 1.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3164-16

 
ricca chemical company
RI316416
Poly natural 500mL Each for $107.48

3164-32

 
ricca chemical company
RI316432
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear blue
Physical Form Liquid
Quantity 500mL
Concentration or Composition Notes 5ppm F-
Equivalence 1 mL = 0.005 mg F-
Chemical Name or Material Fluoride Standard
CAS Max % 1.68
CAS 7647-14-5
CAS Min % 1.56
CAS Max % 1.63
CAS 13291-61-7
CAS Min % 0.11
CAS Max % 0.11
CAS 7681-49-4
CAS Min % 4.0E-4
CAS Max % 4.0E-4
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 1310-73-2
Packaging Poly natural
CAS Min % 1.92
CAS Max % 2.0
CAS 64-19-7
CAS Min % 1.61