missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 1mL = 0.005mg F in H2O, Ricca Chemical
Manufacturer: Ricca Chemical Company RI316916
Specifications
Fluoride Standard | |
98.0,0.0010 | |
FNa | |
[F-].[Na+] | |
41.988 | |
CHEBI:28741 | |
Poly natural | |
Colorless |
7681-49-4,7732-18-5 | |
100.0,0.0010 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium;fluoride | |
5235 | |
Liquid | |
1 mL = 0.005 mg F | |
500mL |
Chemical Identifiers
7681-49-4 | |
41.988 | |
5235 | |
sodium;fluoride |
FNa | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
CHEBI:28741 | |
[F-].[Na+] |