missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 2 ppm F- (Premixed with TISAB/Color Coded Red), Ricca Chemical
Supplier: Ricca Chemical Company 316316
Specifications
Fluoride Standard | |
Natural Poly Bottle | |
93.0,1.94,1.63,1.58,0.11,0.01 | |
Red | |
1 mL = 0.002 mg F- |
7681-49-4,1310-73-2,64-19-7,7647-14-5,13291-61-7,25956-17-6, 7732-18-5 | |
500 mL | |
97.0,2.02,1.69,1.64,0.11,0.01 | |
2ppm F- | |
Liquid |