missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoride Standard, 1mL = 0.002mg F in H2O, Ricca Chemical

$85.30 - $85.30

Chemical Identifiers

CAS 7732-18-5,7681-49-4
CAS Min % 98.0,4.0E-4
CAS Max % 100.0,4.0E-4
Physical Form Liquid
Chemical Name or Material Fluoride Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
3167-16 SDS Ricca Chemical Company
RI316716
Poly natural 500mL Each for $85.30
3167-32 SDS Ricca Chemical Company
RI316732
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-49-4
100.0,4.0E-4
Fluoride Standard
98.0,4.0E-4
Liquid

Specifications

7732-18-5,7681-49-4
100.0,4.0E-4
Poly natural
Clear colorless
Fluoride Standard
98.0,4.0E-4
Liquid
1 mL = 0.002 mg F
500mL