missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoride Standard, 1 ppm F- (Premixed with TISAB/Color Coded Green), Ricca Chemical

Manufacturer:  Ricca Chemical Company 316232

 View more versions of this product

Catalog No. 3162-32



Description

Description

Specifications

Specifications

0.001mg/mL, 1ppm
7681-49-4,1310-73-2,64-19-7,7647-14-5,13291-61-7,3844-45-9
97.0,2.03,1.7,1.65,0.11,1.0E-4
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium;fluoride
5235
Liquid
1mL = 0.001mg F
1 L
Fluoride Standard
93.0,1.95,1.63,1.58,0.11,1.0E-4
FNa
[F-].[Na+]
41.988
CHEBI:28741
Natural Poly Bottle
Green

Chemical Identifiers

7681-49-4
41.988
5235
sodium;fluoride
FNa
PUZPDOWCWNUUKD-UHFFFAOYSA-M
CHEBI:28741
[F-].[Na+]
SDS
Documents
Product Certifications
Promotions

Promotions