missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoride Standard, Certified, 1000ppm ±10ppm (1mL = 1mg F-), LabChem™

Manufacturer:  LabChem LC146002

 View more versions of this product

Catalog No. LC146002



Specifications

Specifications

Fluoride Standard
7681-49-4,7732-18-5
FNa
1000ppm ± 10ppm
[F-].[Na+]
41.988
CHEBI:28741
Liquid
Certified
Soluble in water
1L
1000ppm (1mL = 1mg F-)
99.78,0.22
NaF
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium;fluoride
5235
41.99
Passes Test
Poly Bottle
Colorless

Chemical Identifiers

7681-49-4
41.988
5235
sodium;fluoride
FNa
PUZPDOWCWNUUKD-UHFFFAOYSA-M
CHEBI:28741
[F-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

Documents
Promotions

Promotions