missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluorescein, Sodium Salt, 40% min., EMD Millipore

Manufacturer:  Millipore Sigma FX03255

 View more versions of this product

Catalog No. MFX03255



Specifications

Specifications

Fluorescein, Sodium Salt
Orange-Red
Indicator
C20H10Na2O5
376.27
EP/Reagent
600kg/m3
125g
518-47-8
Acid Yellow 73
Powder

Chemical Identifiers

518-47-8
C20H10Na2O5
Acid Yellow 73
Safety and Handling

Safety and Handling

Recommended Storage : Store at +5°C to +30°C

Documents