missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluorescein, Sodium Salt, 40% min., EMD Millipore

$276.09 - $552.17

Chemical Identifiers

CAS 518-47-8
Physical Form Powder
Molecular Formula C20H10Na2O5
Formula Weight 376.27
Synonym Acid Yellow 73
Chemical Name or Material Fluorescein, Sodium Salt, Indicator
Grade EP/Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MFX03251 Emd Millipore Corporation
FX03251
500g Each for $552.17
MFX03255 Emd Millipore Corporation
FX03255
125g Each for $276.09
Specifications Description & Specifications

Chemical Identifiers

518-47-8
C20H10Na2O5
Acid Yellow 73
EP/Reagent Grade
Powder
376.27
Fluorescein, Sodium Salt, Indicator

Specifications

Orange-red
600kg/m3
Powder
376.27
Fluorescein, Sodium Salt, Indicator
500g
518-47-8
C20H10Na2O5
Acid Yellow 73
EP/Reagent Grade

Safety and Handling

Recommended Storage : Store at +5°C to +30°C