Please login to your online account to display your discounted pricing

Fluorescein, Laser Grade 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Fluorescein
Name Note 99%
CAS 2321-07-5
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C20H12O5
Formula Weight 332.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC410620010

  SDS
fisher scientific acros
AC410620010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 332.3
Quantity 1g
Color Orange to red
Merck Index 15, 4192
Beilstein 19, 222
Purity Grade Pure
RTECS Number LM5075000
TSCA TSCA
EINECS Number 219-031-8
Infrared Spectrum Authentic
Lambda Max 498nm (in basic MeOH)
Molar Absorption Coefficient 63900L/(mole.cm) (lambda max.)
Fluorescence Max 518nm (in basic CH3OH)
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Optimum Lasing 545nm max. (in basic CH3OH)
Suitability (for use as a laser dye) passes
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
Chemical Name or Material Fluorescein
Molecular Formula C20H12O5
Name Note 99%
Assay Percent Range 99%
CAS 2321-07-5
Safety and Handling

Warning

GHS P Statement:
Wear eye protection/face protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.