Please login to your online account to display your discounted pricing

Fluorescein isothiocyanate (FITC) Mouse, APC, Clone: NAWESLEE, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $158.05

Specifications

Antigen Fluorescein isothiocyanate (FITC)
Clone NAWESLEE
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123195

 
life technologies
LS17769180
25μg Each for $71.05

501123196

 
life technologies
LS17769182
100μg Each for $158.05
Description & Specifications

Specifications

Antigen Fluorescein isothiocyanate (FITC)
Clone NAWESLEE
Host Species Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This NAWESLEE monoclonal antibody reacts to fluorescein isothyocianate (FITC), a derivative of fluorescein commonly used in flow cytometry and fluorescent microscopy. FITC-9 can be used for the separation of cells labeled with FITC-conjugated antibodies or for staining. Please note that in some instances the FITC signal can be quenched by the anti-FITC antibody; therefore, this phenomenon should be evaluated empirically.