missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Fluorescein, Free Acid, Reag. Ph. Eur., Honeywell™ Fluka™

Reag. Ph. Eur., free acid

$142.58 - $142.58

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046133 View Documents Honeywell Fluka™
32615100G
100g Plastic Bottle Each for $142.58
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

2321-07-5
332.31
GNBHRKFJIUUOQI-UHFFFAOYSA-N
CHEBI:31624
C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
C20H12O5
MFCD00005050
16850
3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one

Specifications

2321-07-5
C20H12O5
NONH for all modes of transport
≤0.25% (as SO4)
C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
Yes
16850
332.31g/mol
Plastic Bottle
0 to 0.25%
Acid Yellow 73
Brown
Fluorescein, Reag. Ph. Eur.
∽1000kg/m3
MFCD00005050
94324
GNBHRKFJIUUOQI-UHFFFAOYSA-N
3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
332.31
CHEBI:31624
Odourless
Passes test for UV spectrum
Solid
314°C to 316°C
100g
Safety and Handling

Safety and Handling

P305 + P351 + P338

H319

EINECSNumber : 219-031-8

RTECSNumber : LM5075000