Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluorescein diacetate, 97%

$186.84 - $461.07

Chemical Identifiers

Chemical Name or Material Fluorescein diacetate
CAS 596-09-8
Molecular Formula C24H16O7
Formula Weight 416.39
MDL Number MFCD00005062
Beilstein 62575
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2446614

  SDS
alfa aesar™
B2446614
25g Each for $186.84

AAB2446622

  SDS
alfa aesar™
B2446622
100g Each for $461.07
Description & Specifications

Specifications

Formula Weight 416.39
Assay Percent Range 0.97
CAS 596-09-8
TSCA TSCA
Beilstein 62575
Melting Point 200° to 203°C
Quantity 25g
Assay 97%
Molecular Formula C24H16O7
MDL Number MFCD00005062
Chemical Name or Material Fluorescein diacetate

Applications
Fluorescein diacetate stains living cells green, and is a substrate for esterases. It is suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes. It is used in studies on intracellular interactions and membrane permeability.

Solubility
Soluble in chloroform, DMSO, ethanol, and water.

Notes
Store at -20°C. Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.