missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluorescamine [for HPLC Labeling], TCI America™

Manufacturer:  TCI America F01921G

 View more versions of this product

Catalog No. F01921G



Specifications

Specifications

Fluorescamine [for HPLC Labeling]
155°C
38183-12-9
MFCD00005928
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
37927
Crystal-Powder at 20°C
Undesignated
1g
C17H10O4
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, fluorescamine, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
C1=CC=C(C=C1)C2=COC3(C2=O)C4=CC=CC=C4C(=O)O3
278.263
278.26

Chemical Identifiers

38183-12-9
278.263
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
37927
C1=CC=C(C=C1)C2=COC3(C2=O)C4=CC=CC=C4C(=O)O3
C17H10O4
MFCD00005928
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, fluorescamine, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
Safety and Handling

Safety and Handling

TSCA : No

Documents